Anti-GOLM1 Goat Polyclonal Antibody
Catalog # PRSI45-688
Supplier: ProSci Inc.
Specifications
- Antigen name:Golgi Membrane Protein 1
- Antigen symbol:GOLM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:GP73|FLJ22634|golgi membrane protein GP73|golgi membrane protein 1|C9orf155|GOLPH2|GOLM1|FLJ23608|golgi phosphoprotein 2|bA379P1.3|golgi protein|PSEC0257|RP11-379P1.3|73-kD
- Concentration:500 ug/mL
- Immunogen:GOLPH2 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of GOLPH2.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
GOLPH2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
GOLpH 2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GOLM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human