Anti-LAT Goat Polyclonal Antibody
Catalog # PRSI45-080
Supplier: ProSci Inc.
Specifications
- Antigen name:Linker for activation of T-cells Family member 1
- Antigen symbol:LAT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:36 kDa phospho-tyrosine adapter protein|pp36|p36-38|LAT|Linker for activation of T-cells family member 1
- Concentration:500 ug/mL
- Immunogen:LAT antibody was raised against a 12 amino acid synthetic peptide near the C-Terminus of LAT.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
LAT antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
LAT antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: LAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat