Anti-SOX21 Goat Polyclonal Antibody
Catalog # PRSI42-567
Supplier: ProSci Inc.
Specifications
- Antigen name:SRY (sex determining region Y)-box 21
- Antigen symbol:SOX21
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:ALS22|TUBA1|H2-ALPHA
- Concentration:500 ug/mL
- Immunogen:SOX21 antibody was raised against a 13 amino acid synthetic peptide near the internal region of SOX21.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
SOX21 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
SOX21 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SOX21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse