Anti-GNB2L1 Goat Polyclonal Antibody
Catalog # PRSI42-371
Supplier: ProSci Inc.
Specifications
- Antigen name:Guanine Nucleotide Binding Protein (G Protein), Beta Polypeptide 2-like 1
- Antigen symbol:GNB2L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:H12.3|Receptor for activated C kinase|P205|Gnb2-rs1|Receptor of activated protein kinase C 1|PIG21|Guanine nucleotide-binding protein beta subunit-like protein|HLC-7|RACK1
- Concentration:500 ug/mL
- Immunogen:GNB2L1 antibody was raised against a amino acid synthetic peptide near the internal region of GNB2L1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
GNB2L1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
GNB2L1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GNB2L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat