Specifications
- Antigen name:Sodium/phosphate cotransporter
- Antigen symbol:NPT
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Immunogen:Ni-NTA purified truncated recombinant NPT expressed in E. Coli strain BL21 (DE3).
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Neomycin phosphotransferase II (nptII) gene is used in selection of transformed organisms. It was initially isolated from the transposon Tn5 that was present in the bacterium strain Escherichia coli K12. The gene codes for the aminoglycoside 3'-phosphotransferase (denoted aph(3')-II or NPTII) enzyme. NPTII is probably the most widely used selectable marker for plant transformation. It is also used in gene expression and regulation studies in different organisms in part because N-terminal fusions can be constructed that retain enzymatic activity. In animal cells, G418 and neomycin are used as selectable agents.
Antibody is purified by protein A affinity chromatography.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: NPT
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human