Anti-TM9SF1 Rabbit Polyclonal Antibody
Catalog # PRSI31-343
Supplier: ProSci Inc.
Specifications
- Antigen name:Transmembrane 9 Superfamily Member 1
- Antigen symbol:TM9SF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Zebrafish,Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:HMP 70|MP70|Multispanning membrane protein 70kD|MP 70|Transmembrane 9 superfamily member 1|TM9S1_HUMAN.|Transmembrane protein 9 superfamily member 1|HMP70
- Immunogen:A synthetic peptide corresponding a region of human TM9SF1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
TM9SF1 may function as channel, small molecule transporter or receptor.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TM9SF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Zebrafish