Anti-GDE1 Rabbit Polyclonal Antibody
Catalog # PRSI30-276
Supplier: ProSci Inc.
Specifications
- Antigen name:Glycerophosphodiester Phosphodiesterase 1
- Antigen symbol:GDE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Membrane Interacting Protein Of RGS16|MIR16|EC 3.1.4.44
- Immunogen:A synthetic peptide corresponding a region of human GDE1.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
GDE1 has glycerophosphoinositol phosphodiesterase activity. It has little or no activity towards glycerophosphocholine. GDE1 activity can be modulated by G-protein signaling pathways.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GDE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog