Anti-SLC35F2 Rabbit Polyclonal Antibody
PRSI29-930
- Antigen name:Solute Carrier Family 35 Member F2
- Antigen symbol:SLC35F2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:HSNOV1
- Immunogen:A synthetic peptide corresponding a region of human SLC35F2.
- Size:100 µg
- Pk:100 µG
SLC35F2 is a putative solute transporter.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC35F2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human