Anti-KCNN2 Rabbit Polyclonal Antibody
Product Details & Documents
Action potentials in vertebrate neurons are followed by an afterhyperpolarization (AHP) that may persist for several seconds and may have profound consequences for the firing pattern of the neuron. Each component of the AHP is kinetically distinct and is mediated by different calcium-activated potassium channels. The protein encoded by KCNN2 is activated before membrane hyperpolarization and is thought to regulate neuronal excitability by contributing to the slow component of synaptic AHP. The encoded protein is an integral membrane protein that forms a voltage-independent calcium-activated channel with three other calmodulin-binding subunits. KCNN2 is a member of the KCNN family of potassium channel genes.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: KCNN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Documents
10101-076.pdf
Datasheet
Specifications
- Size:100 µg
- Western blot:Yes
- Cat. No.:PRSI28-239
- Clonality:Polyclonal
- Reactivity:Human
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human KCNN2.
- Host:Rabbit
- Conjugation:Unconjugated
- Antigen symbol:KCNN2
- Antigen name:Potassium intermediate/small conductance calcium-activated channel, subfamily N, member 2
- ELISA:Yes
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



