Order Entry
Finland
ContactUsLinkComponent
Anti-PAX3 Rabbit Polyclonal Antibody
Anti-PAX3 Rabbit Polyclonal Antibody
  PRSI27-599
undefined
Anti-PAX3 Rabbit Polyclonal Antibody
  PRSI27-599

 

  • Antigen name:
    Paired Box 3
  • Antigen symbol:
    PAX3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    CDHS|paired box protein Pax-3|WS3|WS1|paired domain gene HuP2|paired domain gene 3|HUP2|paired box homeotic gene 3
  • Immunogen:
    A synthetic peptide corresponding a region of human PAX3.
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

PAX3 is a member of the paired box (PAX) family of transcription factors. Members of the PAX family typically contain a paired box domain and a paired-type homeodomain. These genes play critical roles during fetal development. Mutations in paired box gene 3 are associated with Waardenburg syndrome, craniofacial-deafness-hand syndrome, and alveolar rhabdomyosarcoma. The translocation t (2;13) (q35;q14), which represents a fusion between PAX3 and the forkhead gene, is a frequent finding in alveolar rhabdomyosarcoma.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PAX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat