Order Entry
Finland
ContactUsLinkComponent
 
Anti-FOXD2 Rabbit Polyclonal Antibody
  PRSI27-402
undefined
Anti-FOXD2 Rabbit Polyclonal Antibody
  PRSI27-402

 

  • Antigen name:
    Forkhead box D2
  • Antigen symbol:
    FOXD2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    FREAC-9|FKHL17|FREAC9
  • Immunogen:
    A synthetic peptide corresponding a region of human FOXD2.
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

FOXD2 belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FOXD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human