Anti-SLC10A4 Rabbit Polyclonal Antibody
PRSI25-929
- Antigen name:Solute Carrier Family 10 (sodium/bile acid cotransporter Family) Member A4
- Antigen symbol:SLC10A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human SLC10A4.
- Size:50 µg
- Pk:50 µG
SLC15A4 is the proton oligopeptide cotransporter. It transports free histidine and certain di- and tripeptides.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC10A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human