Order Entry
Finland
ContactUsLinkComponent
Anti-LZTR1 Rabbit Polyclonal Antibody
Anti-LZTR1 Rabbit Polyclonal Antibody
Catalog # PRSI5621
Supplier:  ProSci Inc.
undefined
Anti-LZTR1 Rabbit Polyclonal Antibody
Catalog # PRSI5621
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Leucine Zipper-like Transcription Regulator 1
  • Antigen symbol:
    LZTR1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    Leucine-zipper-like transcriptional regulator 1|LZTR1|TCFL2|SWNTS2|BTBD29|LZTR-1
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    LZTR1 antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human LZTR1.
  • Tested applications:
    E, WB, IHC
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

LZTR1 Antibody: LZTR1, a member of the BTB-kelch superfamily, was initially described as a putative transcriptional regulator based on weak homology to members of the basic leucine zipper-like family, the encoded protein subsequently has been shown to localize exclusively to the Golgi network where it may help stabilize the Golgi complex. Deletion of this gene may be associated with DiGeorge syndrome, a developmental field defect involving the third and fourth pharyngeal pouches, causing the absence of thymus and parathyroid glands, congenital cardiac abnormalities and facial dysmorphism. LZTR1 is tyrosine phosphorylated and subsequently degraded upon induction of apoptosis.

Type:
Antigen: LZTR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat