Anti-TWA1 Rabbit Polyclonal Antibody
Catalog # PRSI5305
Supplier: ProSci Inc.
Specifications
- Antigen name:GID Complex Subunit 8 Homolog
- Antigen symbol:TWA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:TWA1|C20orf11|Chromosome 20 Open Reading Frame 11|GID Complex Subunit 8|Two Hybrid Associated Protein No. 1 With RanBPM
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:TWA1 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human TWA1.
- Tested applications:E, WB, IHC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
TWA1 Antibody: TWA1 was identified through a two hybrid-associated protein screen with RanBPM. TWA1 is well conserved through evolution and is localized within the nucleus. It interacts with RanBP9 and comprises a protein complex with RanBPM and Muskelin. TWA1 was found to possess the LisH-CTLH motif which is detected in proteins involved in microtubule dynamics, cell migration, nucleokinesis and chromosome segregation. These functions overlap with functions suggested for the RanGTPase cycle. Recent findings suggested that there is an as yet uncovered function of the RanGTPase cycle.
Type:
Antigen: TWA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat