0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Standards
Organic Standards
Supelco® Drug and metabolite standards, L-Adrenaline, LoGiCal®
Supelco® Drug and metabolite standards, L-Adrenaline, LoGiCal®
PROOLGCFOR0614.02
:
LGC Standards PROMOCHEM
:
Supelco® Drug and metabolite standards, L-Adrenaline, LoGiCal®
PROOLGCFOR0614.02
:
LGC Standards PROMOCHEM
:
Pk:
10 mg
Standards:
Reference standard
CAS No.:
51-43-4
Description:
L-Adrenaline
Formula:
C₉H₁₃NO₃
Description:
L-Adrenaline