0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Standards
Organic Standards
Drug and metabolite standards, digoxin, LoGiCal®
Drug and metabolite standards, digoxin, LoGiCal®
PROOLGCFOR0212.00
:
LGC Standards PROMOCHEM
:
Drug and metabolite standards, digoxin, LoGiCal®
PROOLGCFOR0212.00
:
LGC Standards PROMOCHEM
:
Pk:
10 mg
Standards:
Reference standard
CAS No.:
20830-75-5
Description:
Digoxin
Formula:
C₄₁H₆₄O₁₄
Description:
Digoxin