Drug and metabolite standards, androstenedione, LoGiCal
Catalog # PROOLGCFOR0323.01
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:10 mg
- Standards:Reference standard
- CAS No.:63-05-8
- Description:Androstenedione
- Formula:C₁₉H₂₆O₂
- Description:Androstenedione