Order Entry
Finland
Orders LinkContactUsLinkComponent
Carteolol, Reference Standards

Carteolol, Reference Standards

Supplier:   USP
Carteolol, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1166400.
  • 1096757.
  • 1096859.
  • Description:
  • Dehydrocarteolol Hydrochloride
  • Carteolol Hydrochloride
  • Carteolol
  • Synonym(s):
  • 5-[3-(tert-Butylamino)-2-hydroxypropoxy]quinolin-2(1H)-one hydrochloride
  • 5-(2-Hydroxy-3-methoxypropoxy)-3,4-dihydroquinolin-2(1H)-one
  • Physical form:
  • Solid, powder
  • CAS No.:
  • 53371-79-2
  • 51781-21-6
  • Formula:
  • C₁₆H₂₄N₂O₃·HCl
  • C₁₃H₁₇NO₄
  • Storage conditions:
  • Refigerator

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1166400.
    Dehydrocarteolol Hydrochloride
    Dehydrocarteolol Hydrochloride
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 744,00 €
    744,00 €
    1 * 100 mg
       744,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1166400.
    Description
    Dehydrocarteolol Hydrochloride
    Synonym(s)
    5-[3-(tert-Butylamino)-2-hydroxypropoxy]quinolin-2(1H)-one hydrochloride
    Physical form
    CAS No.
    53371-79-2
    Formula
    Storage conditions
  • 1096757.
    Carteolol Hydrochloride
    Carteolol Hydrochloride
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 395,00 €
    395,00 €
    1 * 200 mg
       395,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1096757.
    Description
    Carteolol Hydrochloride
    Synonym(s)
    Physical form
    CAS No.
    51781-21-6
    Formula
    C₁₆H₂₄N₂O₃·HCl
    Storage conditions
  • 1096859.
    Carteolol
    Carteolol
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 780,00 €
    1 780,00 €
    1 * 25 mg
       1 780,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1096859.
    Description
    Carteolol
    Synonym(s)
    5-(2-Hydroxy-3-methoxypropoxy)-3,4-dihydroquinolin-2(1H)-one
    Physical form
    Solid, powder
    CAS No.
    Formula
    C₁₃H₁₇NO₄
    Storage conditions
    Refigerator
Suositukset

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.