Order Entry
Finland
ContactUsLinkComponent
Sodium carbonate - acidimetric standard
Sodium carbonate - acidimetric standard
Catalog # PROONIST-351A
CAS Number:  
Sodium carbonate - acidimetric standard
Catalog # PROONIST-351A
CAS Number:  
Specifications
  • Pk:
    50 g
  • Description:
    NIST-351A sodium carbonate - acidimetric standard
  • Element:
    Sodium
  • Symbol:
    Na
Specifications
About this item
These standards are intended for use as a acidimetric standard.
The SRM consists of highly purified sodium carbonate in a 50 g unit.
Certifications: Traceable to NIST and SRM.