Order Entry
Finland
ContactUsLinkComponent
Spigot with screw caps for jerricans
Spigot with screw caps for jerricans
Catalog # HUEN550500
Supplier:  hünersdorff GmbH
Spigot with screw caps for jerricans
Catalog # HUEN550500
Supplier:  hünersdorff GmbH
Specifications
  • Description:
    Spigot PROFI, natural, with screw cap
  • Pk:
    5
Specifications
About this item
Spigots PROFI with red DIN 60/61 screw cap is ideal for controlled emptying of liquids from industrial jerrycans.
  • Resistant to acids, alkalis and other chemicals
  • Pressure resistance up to 0,4 bar