Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Tinidazole, Reference Standards

Supplier:   USP
Tinidazole, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1667541.
  • 1667530.
  • 1667520.
  • Description:
  • Tinidazole Related Compound B
  • Tinidazole Related Compound A
  • Tinidazole
  • Synonym(s):
  • 1-(2-ethyl-sulfonylethyl)-2-methyl-4-nitroimidazole
  • 2-methyl-5-nitroimidazole
  • CAS No.:
  • 25459-12-5
  • 696-23-1
  • 19387-91-8
  • Formula:
  • C₈H₁₃N₃O₄S
  • C₄H₅N₃O₂
  • C₈H₁₃N₃O₄S

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1667541.
    Tinidazole Related Compound B
    Tinidazole Related Compound B
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 300,00 €
    1 300,00 €
    1 * 20 mg
       1 300,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1667541.
    Description
    Tinidazole Related Compound B
    Synonym(s)
    1-(2-ethyl-sulfonylethyl)-2-methyl-4-nitroimidazole
    CAS No.
    25459-12-5
    Formula
    C₈H₁₃N₃O₄S
  • 1667530.
    Tinidazole Related Compound A
    Tinidazole Related Compound A
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 1 280,00 €
    1 280,00 €
    1 * 100 mg
       1 280,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1667530.
    Description
    Tinidazole Related Compound A
    Synonym(s)
    2-methyl-5-nitroimidazole
    CAS No.
    696-23-1
    Formula
    C₄H₅N₃O₂
  • 1667520.
    Tinidazole
    Tinidazole
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 365,00 €
    365,00 €
    1 * 200 mg
       365,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1667520.
    Description
    Tinidazole
    Synonym(s)
    CAS No.
    19387-91-8
    Formula
    C₈H₁₃N₃O₄S