Order Entry
Finland
Orders LinkContactUsLinkComponent
Tacrolimus, Reference Standards

Tacrolimus, Reference Standards

Supplier:   USP
Tacrolimus, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1642857.
  • 1642802.
  • Description:
  • Tacrolimus System Suitability Mixture
  • Tacrolimus
  • CAS No.:
  • 109581-93-3
  • Formula:
  • C₄₄H₇₁NO₁₃
  • UN No.:
  • 2811
  • 2811

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1642857.
    Tacrolimus System Suitability Mixture
    Tacrolimus System Suitability Mixture
    USP reference standard
    Vial
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 1 690,00 €
    1 690,00 €
    1 * 10 mg
       1 690,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1642857.
    Description
    Tacrolimus System Suitability Mixture
    CAS No.
    Formula
    UN No.
    2811
  • 1642802.
    Tacrolimus
    Tacrolimus
    USP reference standard
    Vial
    150 mg
    Log in for availability
    Log in for availability
    1 * 150 mg
    1 * 150 mg - 1 450,00 €
    1 450,00 €
    1 * 150 mg
       1 450,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1642802.
    Description
    Tacrolimus
    CAS No.
    109581-93-3
    Formula
    C₄₄H₇₁NO₁₃
    UN No.
    2811