Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Oleyl Alcohol, Reference Standard

Oleyl Alcohol, USP reference standard, 5 x 50 mg, Ampule, 5
Catalog # 1478185.
Supplier:   USP
Oleyl Alcohol, Reference Standard
Catalog # 1478185.
Supplier:  USP
CAS Number:  
Oleyl Alcohol, Reference Standard

Oleyl Alcohol, USP reference standard, 5 x 50 mg, Ampule, 5

CAS Number:  
Price
443,00 €
Log in for availability
1 * 5 Items
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

Specifications

Specifications

  • Pack type:
    Ampule
  • Pk:
    5
  • Standards:
    USP reference standard
  • Description:
    Oleyl Alcohol
  • Pk Info:
    5 x 50 mg
  • Cat. No.:
    1478185.
  • Formula:
    C₁₈H₃₆O
  • Description:
    Oleyl Alcohol
  • CAS No.:
    143-28-2

Specifications