Order Entry
Finland
ContactUsLinkComponent
Low alloy steel standard
Low alloy steel standard
Catalog # PROONIST-1764A
CAS Number:  
Low alloy steel standard
Catalog # PROONIST-1764A
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    NIST-1764A, low alloy steel
Specifications
About this item
This reference material is intended for use in the evaluation of chemical and instrumental analysis and in calibration of those methods.
  • 22 analytes
A unit consists of a disk approximately 34 mm in diameter and 19 mm thick.
Certifications: Traceable to NIST and SRM.