Order Entry
Finland
ContactUsLinkComponent
Impurity and API standard, tocopheryl acetate, all-rac-alpha-
Impurity and API standard, tocopheryl acetate, all-rac-alpha-
  PROOMM1381.00
 :  
Impurity and API standard, tocopheryl acetate, all-rac-alpha-
  PROOMM1381.00
 :  

 

  • Pk:
    250 mg
  • Standards:
    Reference standards
  • CAS No.:
    7695-91-2
  • Description:
    all-rac-alpha-tocopheryl acetate
  • Formula:
    C₃₁H₅₂O₃
  • Description:
    all-rac-alpha-tocopheryl acetate