Order Entry
Finland
ContactUsLinkComponent
 
Impurity and API standard, dacarbazine
  PROOMM1289.00
 :  
Impurity and API standard, dacarbazine
  PROOMM1289.00
 :  

 

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    4342-03-4
  • Description:
    Dacarbazine
  • Formula:
    C₆H₁₀N₆O
  • Description:
    Dacarbazine