Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Impurity and API standard, synephrine
Impurity and API standard, synephrine
Impurity and API standard, synephrine
Catalog # PROOMM0813.00
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Expand image
Impurity and API standard, synephrine
Catalog # PROOMM0813.00
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Specifications
Pk:
1
Standards:
Reference standards
CAS No.:
94-07-5
Description:
Synephrine
Formula:
C₉H₁₃NO₂
Description:
Synephrine
Specifications
About this item
Product Details
Product Details