Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Impurity and API standard, dibenzocycloheptene
Impurity and API standard, dibenzocycloheptene
Impurity and API standard, dibenzocycloheptene
PROOMM0609.01
:
LGC Standards PROMOCHEM
:
Impurity and API standard, dibenzocycloheptene
PROOMM0609.01
:
LGC Standards PROMOCHEM
:
Pk:
50 mg
Standards:
Reference standards
CAS No.:
256-81-5
Description:
5H-Dibenzo[a,d][7]annulene
Formula:
C₁₅H₁₂
Description:
5H-Dibenzo[a,d][7]annulene