Order Entry
Finland
ContactUsLinkComponent
 
Impurity and API standard, hyoscine N-oxide hydrobromide
Catalog # PROOMM0546.07
CAS Number:  
Impurity and API standard, hyoscine N-oxide hydrobromide
Catalog # PROOMM0546.07
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    6106-81-6
  • Description:
    Hyoscine N-oxide hydrobromide
  • Formula:
    C₁₇H₂₁NO₅.BrH
  • Description:
    Hyoscine N-oxide hydrobromide

Specifications

About this item