Combustion trays, ceramic, AL24
33187.KF
- Capacity:240 ml
- Seq:01
- W×D×H:145 x 85 x 30 mm
- Pk:1
Pure Al₂O₃ Aluminium oxide AL24.
Slightly porous, good resistance to thermal shock, extremely good creep strength.
Pure Al₂O₃ Aluminium oxide AL24.
Slightly porous, good resistance to thermal shock, extremely good creep strength.