Order Entry
Finland
ContactUsLinkComponent
VWR® pH CO 1030, pH/Conductivity Tester
VWR® pH CO 1030, pH/Conductivity Tester
Catalog # 665-0480
Supplier:  VWR Collection
undefined
VWR® pH CO 1030, pH/Conductivity Tester
Catalog # 665-0480
Supplier:  VWR Collection

Specifications

  • Temperature range:
    −5,0...+60,0 °C
  • Description:
    pH/conductivity tester, pH/CO1030
  • Model:
    pH/CO1030
  • Temperature accuracy:
    ±0,5 °C
  • Temperature resolution:
    0,1 °C
  • Automatic buffer recognition:
    pH 7,00/4,01/10,01 or pH 6,86/4,00/9,18
  • Compliance:
    IP 67
  • Conductivity accuracy:
    ±1% FS
  • Conductivity range:
    0 µS/cm - 20,00 mS/cm
  • Conductivity resolution:
    Minimum 1 µS/cm
  • Data logging capacity:
    50 data sets
  • Power supply:
    4× LR44 batteries
  • Reference temperature:
    15,0 - 25,0 °C
  • Salinity accuracy:
    ±1% FS
  • Salinity range:
    0,0 - 10,0 ppt
  • Salinity resolution:
    0,1 ppt
  • TDS accuracy:
    ±1% FS
  • TDS conversion factor:
    0,30 - 1,00 (default: 0,65)
  • TDS range:
    0 mg/L - 20,00 g/L
  • TDS resolution:
    Minimum 1 mg/L
  • Temperature compensation:
    Automatic (−5,0...+60,0 °C)
  • Weight:
    0,095 kg
  • W×D×H:
    44×25×200 mm
  • pH accuracy:
    ±0,2
  • pH calibration:
    1-, 2- or 3-point
  • pH range:
    0,00 - 14,00
  • pH resolution:
    0,01
  • Pk:
    1

Specifications

About this item

This tester is a convenient pen instrument for measuring the following water quality parameters: pH, conductivity, TDS, salinity, and temperature.

  • Graphic LCD, IP 67 waterproof housing
  • Replaceable electrode
  • Automatic temperature compensation; automatic calibration and buffer recognition
  • HOLD function, battery indicator, automatic power off function after 10 minutes of non use
  • Non volatile memory stores up to 50 sets of data (pH/conductivity/TDS/salinity and temperature) even if power is lost
  • Belt clip for easy storage when not in use

Warranty: 1 year (instrument)/6 months (electrode)

Delivery: Supplied with pre-installed pH/conductivity electrode assembly and electrode cap, manual and set of 4 batteries.