VWR® Centrifuge Tubes, Metal-Free
Supplier: VWR Collection
Certificates
About this item
PP, conical, with natural HDPE flat cap with integral seal.
- Available in two sizes with natural flat cap with integral sealing for easy labelling
- Tubes and caps are made of ultra-high purity CFR 21 compliant resins
- Printed graduation marks and large white writing area
- Sustainable design based on virgin resins
For environmental uses such as water analysis and other applications where tubes have the potential to contaminate samples. Tubes are certified free of major critical trace metals to less than 1 ppb.
Certifications: Manufactured in a Class 100000 room environment.
Packaging: Tubes packed in bulk bags are supplied 50 tubes per bag, 10 bags per case. SBS recyclable racked tubes are supplied 25 per rack, 2 racks per pack, 10 packs per case.
Specifications
- Catalog No:
- 525-0463
- 525-0460
- Environmentally Preferable:
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging