Order Entry
Finland
ContactUsLinkComponent
General purpose gloves, Abratex Light Grip
General purpose gloves, Abratex Light Grip
Supplier:  HONEYWELL SAFETY
Certificates
undefined
General purpose gloves, Abratex Light Grip
Supplier:  HONEYWELL SAFETY
Specifications
  • Catalog No:
  • DALLRGT888/8
  • DALLRGT888/10
  • DALLRGT888/7
  • DALLRGT888/9
  • Model:
  • Abratex Light Grip
  • Abratex Light Grip
  • Abratex Light Grip
  • Abratex Light Grip
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Finish:
  • Dot coating on palm
  • Dot coating on palm
  • Dot coating on palm
  • Dot coating on palm
  • Gauge:
  • 13
  • 13
  • 13
  • 13
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (224XX)
  • EN 388:2016 (224XX)
  • EN 388:2016 (224XX)
  • EN 388:2016 (224XX)
Specifications