Order Entry
Finland
ContactUsLinkComponent
VWR® B'Wipes 5000, Cleanroom wipes
VWR® B'Wipes 5000, Cleanroom wipes
Catalog # 115-1902
Supplier:  VWR Collection
undefined
VWR® B'Wipes 5000, Cleanroom wipes
Catalog # 115-1902
Supplier:  VWR Collection

Specifications

  • Packed:
    Stacked, 2×75
  • Description:
    Series 5000 wipes
  • L×W:
    230×230 mm
  • Seq:
    09
  • Pk:
    150

Specifications

About this item

Polyester, sealed-border wipes for use in ISO 3 cleanrooms.

  • Double-knit, polyester wipe with sealed borders (135 g/m²)
  • Laundered, with sealed borders, for low release of fibres even under mechanical stress
  • Good mechanical resistance and absorption capacity
  • Ideal for cleaning heavy soils and disinfecting surfaces in critical areas