Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 72-8074-U500
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Rabbit
- Western blot:Yes
- Epitope:Heavy and light chain
- Cross adsorption:Yes
- Antibody fragment:F(ab')2
- Storage temperature:Store freeze-dried powder at 2-8 °C. Once rehydrated and in solution store at 2-8 °C.
- Size:0.5 mg
- Pk:0,5 mg
Specifications
About this item
Goat anti-Rabbit IgG (H&L), F(ab')2 Fragment - Affinity Pure, HRP Conjugate, min x w/ bovine, human, or mouse IgG or serum proteins
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Rabbit