Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin))
Catalog # 50-8036-U100
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Goat
- Reactivity:Rabbit
- Epitope:Heavy and light chain
- Storage temperature:Store freeze-dried powder at 2-8 °C. Once rehydrated and in solution store at 2-8 °C.
- Pk:100 µG
Specifications
About this item
Goat anti-Rabbit IgG, secondary antibody, conjugated toSureLight® R-Phycoerythrin
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Rabbit