Anti-IgG Sheep Polyclonal Antibody
70-8078-M002
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Sheep
- ImmunoChemistry:Yes
- Reactivity:Rabbit
- Western blot:Yes
- Epitope:Heavy and light chain
- Storage temperature:Store at 2-8 °C
- Pk:2 mg
Sheep anti-Rabbit IgG (H&L) - Affinity Pure
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and light chain
Host: Sheep
Isotype:
Reactivity: Rabbit