Vacuum adapter, ground joint, PTFE, BOLA
Supplier: Bohlender
Certificates
About this item
This vacuum adapter is made of PTFE with O-ring made of FKM for secure placement of standard filter cartridges made of glass or plastic on vessels and round flasks with ground joint.
- Lateral step-type solenoid made of PTFE
- Chemical resistance
- Temperature resistance from –15 to +200 °C
For vacuum filtration, the O-ring at the passageway seals the connection to the filter slide so that even a small vacuum is sufficient.