Urea 98.0-102% USP, Macron Fine Chemicals™
:57-13-6
:
:NH₂CONH₂
:MFCD00008022
:13,09935
:197 °C (1013 hPa)
:132.7 °C
:1.34 g/cm³ (20 °C)
:60.06 g/mol
:Ambient
:200-315-5
:01-2119463277-33
- USP - Assay (NH₂CONH₂):98.0 - 102.0 %
- USP - Alcohol Insoluble Matter:≤0.04 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Residue on Ignition:≤0.1 %
- USP - Organic impurities: Urea related compound A:≤0.1 %
- USP - Organic impurities: Any individual unspecified impurity:≤0.1 %
- USP - Organic impurities: Total impurities:≤2.0 %