Sodium thiosulphate pentahydrate Ph. Eur., Macron Fine Chemicals™
Catalog # P121-33
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:248.19 g/mol
CAS Number:10102-17-7
Formula:Na₂S₂O₃.5H₂O
MDL Number:MFCD00149186