Order Entry
Finland
ContactUsLinkComponent
Sodium hydrogen carbonate 99.7-100.3% (by acidimetry), AR® ACS, Macron Fine Chemicals™
Sodium hydrogen carbonate 99.7-100.3% (by acidimetry), AR® ACS, Macron Fine Chemicals™
Certificates
undefined
Sodium hydrogen carbonate 99.7-100.3% (by acidimetry), AR® ACS, Macron Fine Chemicals™
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    99.7 - 100.3 %
  • ACS - Assay (NaHCO₃) (by acidimetry):
    99.7 - 100.3 %
  • ACS - Ammonium (NH₄):
    ≤5 ppm
  • ACS - Calcium (Ca):
    ≤0.02 %
  • ACS - Chloride (Cl):
    ≤0.003 %
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Insoluble Matter:
    ≤0.015 %
  • ACS - Iron (Fe):
    ≤0.001 %
  • ACS - Magnesium (Mg):
    ≤0.005 %
  • ACS - Phosphate (PO₄):
    ≤0.001 %
  • ACS - Potassium (K):
    ≤0.005 %
  • ACS - Sulfur Compounds (as SO₄):
    ≤0.003 %

Specification Test Results

Related Information