Order Entry
Finland
ContactUsLinkComponent
 
Silver nitrate 99.0-100.5% Ph. Eur., Macron Fine Chemicals™
 
undefined
Silver nitrate 99.0-100.5% Ph. Eur., Macron Fine Chemicals™
 

 

  • Ph. Eur. - Assay:
    99.0 - 100.5%
  • Ph. Eur. - Identification A:
    Passes Test
  • Ph. Eur. - Identification B:
    Passes Test
  • Ph. Eur. - Appearance of Solution:
    Passes Test
  • Ph. Eur. - Acidity or Alkalinity:
    Passes Test
  • Ph. Eur. - Foreign Salts:
    ≤0.3%
  • Ph. Eur. - Aluminium, Lead, Copper and Bismuth:
    Passes Test
  • Clarity and Colour of solution:
    Clear, Colourless
  • Identity A:
    Positive Reaction
  • Identity B:
    Positive Reaction
  • Copper:
    Does not Appear Blue Colour
  • Assay:
    99.8 - 100.5%