Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Propanol
2-Propanol Ph. Eur., Macron Fine Chemicals™
2-Propanol Ph. Eur., Macron Fine Chemicals™
2-Propanol Ph. Eur., Macron Fine Chemicals™
Supplier:
MACRON AVANTOR BRAND
Certificates
Expand image
2-Propanol Ph. Eur., Macron Fine Chemicals™
Supplier:
MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:
67-63-0
:
Formula:
(CH₃)₂CHOH
MDL Number:
MFCD00011674
:
60.1 g/mol
UN:
1219
ADR:
3,II
Related Information
Documentation
Certificates
Search by Lot Number
SDS
sds_MACR7065
Documentation