Order Entry
Finland
ContactUsLinkComponent
Bismuth(III) oxide ≥99.5% (by EDTA titration), BAKER ANALYZED® for electronic ceramics, J.T.Baker®
Bismuth(III) oxide ≥99.5% (by EDTA titration), BAKER ANALYZED® for electronic ceramics, J.T.Baker®
Catalog # 1160-08
Supplier:  Avantor
CAS Number:  
undefined
Bismuth(III) oxide ≥99.5% (by EDTA titration), BAKER ANALYZED® for electronic ceramics, J.T.Baker®
Catalog # 1160-08
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    50 lb
  • Seq:
    7137

Specifications

Specification Test Results

  • For Microelectronic Use:
  • Assay (as Bi₂O₃) (by EDTA titrn):
    ≥99.5 %
  • Chloride (Cl):
    ≤0.001 %
  • Nitrate (NO₃):
    ≤0.005 %
  • Total Sulfur (as SO₄):
    ≤0.002 %
  • Aluminum (Al):
    ≤0.005 %
  • Calcium (Ca):
    ≤0.005 %
  • Copper (Cu):
    ≤0.001 %
  • Iron (Fe):
    ≤0.003 %
  • Lead (Pb):
    ≤0.005 %
  • Silicon (Si):
    ≤0.005 %
  • Sodium (Na):
    ≤0.001 %
  • Average Particle Diameter, µm (APD):
    ≤3
  • Specific Surface Area, m²/g, For Information Only:
    For Information Only
  • Bulk Density (g/cc), For Information Only:
    For Information Only
  • Mesh (Wet Screen Analysis) - On U.S. No. 325 Sieve, For Information Only:
    For Information Only

Specification Test Results

About this item

 :
 :465.96 g/mol
Storage Temperature:Ambient
CAS Number:1304-76-3
Formula:BiO₃
MDL Number:MFCD00003462