Order Entry
Finland
ContactUsLinkComponent
Membrane disc filters, GLA-5000 PVC
Membrane disc filters, GLA-5000 PVC
Certificates
undefined
Membrane disc filters, GLA-5000 PVC
Certificates
Specifications
  • Catalog No:
  • 514-0466
  • 514-0468
  • 514-0467
  • 514-0469
  • Type:
  • GLA-5000
  • GLA-5000
  • GLA-5000, with pads
  • GLA-5000
  • Pore size:
  • 5,00 µm
  • 5,00 µm
  • 5,00 µm
  • 5,00 µm
  • Airflow rate:
  • 53 ml/min/cm² (0,7 bar)
  • 53 ml/min/cm² (0,7 bar)
  • 53 ml/min/cm² (0,7 bar)
  • 53 ml/min/cm² (0,7 bar)
  • Max. temperature:
  • 52 °C
  • 52 °C
  • 52 °C
  • 52 °C
  • Ash content:
  • <1%
  • <1%
  • <1%
  • <1%