Anti-CD300E Mouse Monoclonal Antibody (PE) [clone: UP-H2]
ABNOMAB15414
: Abnova
- Antibody type:Primary
- Antigen name:CD300E molecule
- Antigen symbol:CD300E
- Clonality:Monoclonal
- Clone:UP-H2
- Conjugation:PE (Phycoerythrin)
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:342510
- Storage buffer:In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
- Storage temperature:Store in the dark at 4 °C. Avoid prolonged exposure to light.
- Immunogen:IREM-2-HA-transfected cells.
- Purification:Affinity purification
- Size:100 AS
- Pk:100 Assays
Mouse monoclonal antibody raised against native human CD300E.
Type: Primary
Antigen: CD300E
Clonality: Monoclonal
Clone: UP-H2
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human