One-piece fingertight fittings
Supplier: VICI JOUR RESEARCH
Certificates
About this item
PEEK one-piece fingertight fittings. 10-32 threads, for 1/16" tubing. Fingertight up to 350 bar (5000 psi).
- Excellent chemical resistance
- Machined, not molded
- One-piece – no ferrule required
- Biocompatible