Safety goggles, 2890 series
Supplier: 3M
Certificates
About this item
With a modern slim design, the 2800 series goggles combine performance, comfort and style while providing the wearer with an excellent field of vision. Available with PC or acetate lenses with or without indirect ventilation.
- Easy to adjust strap
- Pivoting ratchet for convenient individual adjustment and fit
- Sealed (111-0493) or indirect ventilation
Weight: 99 g.
EN 166, EN 170
Certifications: EN 166, EN 170