Chemical and cut resistant gloves, Showa 3416
Supplier: Showa
Certificates
About this item
Constructed with a seamless knit liner for maximum comfort and endurance, this fully coated neoprene gauntlet provides ANSI level A4 and EN level 5 cut protection and protection against acids, caustics, solvents, grease and oil.
- Seamless liner made from cut resistant engineered fibre
- Fully coated
- Flexible coating with maximum comfort and protection
- Rough particle finish
- Gauntlet cuff for protection of the forearm
Recommended industries: Transport, steel erecting, mechanical, metallurgy, petrochemical, automotive.
Certifications: EN 388:2016 (3X43E), EN ISO 374-1 Type A (AJKLMT), EN ISO 374-5; Cat. III
Specifications
- Catalog No:
- 112-5784
- 112-5786
- 112-5787
- 112-5785
- Model:
- 3416
- 3416
- 3416
- 3416
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Finish:
- Rough particle finish
- Rough particle finish
- Rough particle finish
- Rough particle finish
- Gauge:
- 13
- 13
- 13
- 13
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN 388:2016 (3X43E), EN ISO 374-1 Type A (AJKLMT), EN ISO 374-5
- EN 388:2016 (3X43E), EN ISO 374-1 Type A (AJKLMT), EN ISO 374-5
- EN 388:2016 (3X43E), EN ISO 374-1 Type A (AJKLMT), EN ISO 374-5
- EN 388:2016 (3X43E), EN ISO 374-1 Type A (AJKLMT), EN ISO 374-5