Order Entry
Finland
ContactUsLinkComponent
Synthetic GLP7-37
Synthetic GLP7-37
Catalog # ROCK000-001-M40
CAS Number:  
Synthetic GLP7-37
Catalog # ROCK000-001-M40
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Conjugation:
    Unconjugated
  • Protein function:
    Growth factor
  • Protein/peptide type:
    Synthetic
  • Protein synonyms:
    GLP-1 analogue|Diabetes research Peptide|Blocking Peptide|Control Peptide|Acylated glucagon-like Peptide-1
  • Protein/peptide name:
    GLP7-37

Specifications

About this item

Liraglutide is a human glucagon-like peptide-1 (GLP-1) agonist. GLP-1 (7-37) activates the GLP-1 cell-surface membrane-bound receptor in pancreatic beta cells and regulates insulin and glucagon secretions. Suitable for use in ELISA, western Blot, dot blot, PCA, and other assays.

  • Lyophilised
  • Purity: >95%
  • Concentration: 1,0 mg/ml by dry weight
  • Store at 2 to 8 °C

Application: Control peptide